Learn More
Aviva Systems Biology Corp USP29 Antibody - N-terminal region (ARP59320_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59320_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against USP29. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human USP29. Alternate names: HOM-TES-84/86, MGC163266, MGC163270ApplicationWB. Species reactivity: Human
Catalog No. 50-286-660