Learn More
Aviva Systems Biology Corp USP3 Antibody - middle region (ARP33681_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33681_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against USP3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human USP3. Alternate names: MGC129878, MGC129879, SIH003, UBPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55116