Learn More
Aviva Systems Biology Corp USP30 Antibody - middle region (ARP50098_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50098_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against USP30. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human USP30. Alternate names: FLJ40511, MGC10702ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-486