Learn More
Medchemexpress LLC HY-141659 5mg , USP30 inhibitor 18 CAS:2242582-40-5 Purity:>98%
SDP
Catalog No. 502259358
Encompass_Preferred
HY-141659 5mg USP30 inhibitor 18 CAS: 2242582-40-5 USP30 inhibitor 18 is a selective USP30 inhibitor with an IC50 of 0.02 μM. USP30 inhibitor 18 increases protein ubiquitination and accelerates mitophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9358 Supplier Medchemexpress LLC Supplier No. HY1416595MG
HY-141659 5mg USP30 inhibitor 18 CAS: 2242582-40-5 USP30 inhibitor 18 is a selective USP30 inhibitor with an IC50 of 0.02 μM. USP30 inhibitor 18 increases protein ubiquitination and accelerates mitophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.