Learn More
Aviva Systems Biology Corp USP35 Antibody - C-terminal region (ARP59325_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59325_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against USP35. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human USP35. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-286-661