Learn More
Aviva Systems Biology Corp USP36 Antibody - N-terminal region (ARP57666_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57666_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against USP36. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human USP36. Alternate names: DUB1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human
Catalog No. 50-014-62920