Learn More
Aviva Systems Biology Corp USP39 Antibody - N-terminal region (ARP38825_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38825_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against USP39. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human USP39. Alternate names: 65K, SAD1, CGI-21, HSPC332, SNRNP65ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56816