Learn More
ABclonal Technology USP47 Rabbit pAb
SDP
Catalog No. 5000105618
Encompass_Preferred
Enables WD40-repeat domain binding activity and thiol-dependent deubiquitinase. Involved in several processes, including monoubiquitinated protein deubiquitination, negative regulation of G2/M transition of mitotic cell cycle, and negative regulation of nitrogen compound metabolic process. Located in cytoplasm. Part of SCF ubiquitin ligase complex.
Catalog No. 50-001-05618 Supplier ABclonal Technology Supplier No. A154611000UL
Enables WD40-repeat domain binding activity and thiol-dependent deubiquitinase. Involved in several processes, including monoubiquitinated protein deubiquitination, negative regulation of G2/M transition of mitotic cell cycle, and negative regulation of nitrogen compound metabolic process. Located in cytoplasm. Part of SCF ubiquitin ligase complex.