Learn More
Aviva Systems Biology Corp USP48 Antibody - middle region (ARP44805_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44805_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against USP48. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human USP48. Alternate names: USP31, RAP1GA1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58901