Learn More
Aviva Systems Biology Corp USP48 Antibody - C-terminal region (ARP44806_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44806_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against USP48. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human USP48. Alternate names: DKFZp762M1713, MGC132556, MGC14879, RAP1GA1, USP31ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58902