Learn More
Aviva Systems Biology Corp USP7 Antibody - middle region (ARP59057_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59057_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against USP7. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human USP7. Alternate names: HAUSP, TEF1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-569