Learn More
Medchemexpress LLC HY-16709 5mg Medchemexpress, USP7-IN-1 CAS:1381291-36-6 Purity:>98%
SDP
Catalog No. 502030732
Encompass_Preferred
Medchemexpress, HY-16709 5mg USP7-IN-1 CAS:1381291-36-6 USP7-IN-1 is a selective and reversible inhibitor of ubiquitin-specific protease 7 (USP7), with an IC50 of 77 μM, and can be used for the research of cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0732 Supplier Medchemexpress LLC Supplier No. HY167095MG
Medchemexpress, HY-16709 5mg USP7-IN-1 CAS:1381291-36-6 USP7-IN-1 is a selective and reversible inhibitor of ubiquitin-specific protease 7 (USP7), with an IC50 of 77 μM, and can be used for the research of cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.