Learn More
Medchemexpress LLC USP7-IN-10 hydrochloride | 2755995-01-6 | 99.3% | 549.51 | 25 MG
SDP
Catalog No. 5000362664
Encompass_Preferred
USP7-IN-10 hydrochloride is a potent ubiquitin-specific protease 7 (USP7) inhibitor with an IC50 of 13.39 nM. This compound targets deubiquitinase and is involved in the cell cycle/DNA damage pathway. It is provided as a light yellow to yellow solid-liquid mixture.
  • Potent ubiquitin-specific protease 7 (USP7) inhibitor
  • IC50 of 13.39 ± 0.04 nM (USP7)
  • Associated with cell cycle/DNA damage pathway
  • Appearance is light yellow to yellow (solid-liquid mixture)
Catalog No. 50-003-62664 Supplier Medchemexpress LLC Supplier No. 25MGHY148481A
USP7-IN-10 hydrochloride is a potent ubiquitin-specific protease 7 (USP7) inhibitor with an IC50 of 13.39 nM. This compound targets deubiquitinase and is involved in the cell cycle/DNA damage pathway. It is provided as a light yellow to yellow solid-liquid mixture.
  • Potent ubiquitin-specific protease 7 (USP7) inhibitor
  • IC50 of 13.39 ± 0.04 nM (USP7)
  • Associated with cell cycle/DNA damage pathway
  • Appearance is light yellow to yellow (solid-liquid mixture)