Learn More
Medchemexpress LLC USP7-IN-10 hydrochloride | 2755995-01-6 | 99.31% | 549.51 | 1 ML
SDP
Catalog No. 5000362083
Encompass_Preferred
USP7-IN-10 hydrochloride (compound 1) is a potent ubiquitin-specific protease 7 (USP7) inhibitor with an IC50 of 13.39 nM.
  • Potent USP7 inhibitor
  • IC50 of 13.39 nM
  • For research use only
Catalog No. 50-003-62083 Supplier Medchemexpress LLC Supplier No. HY148481A10MM/1ML
USP7-IN-10 hydrochloride (compound 1) is a potent ubiquitin-specific protease 7 (USP7) inhibitor with an IC50 of 13.39 nM.
  • Potent USP7 inhibitor
  • IC50 of 13.39 nM
  • For research use only