Learn More
Medchemexpress LLC USP7-IN-3 | 2202738-42-7 | 99.1% | 568.59 | 10 MG
SDP
Catalog No. 5000436063
Encompass_Preferred
USP7-IN-3 (Compound 5) is an effective and selective allosteric inhibitor of ubiquitin-specific protease 7 (USP7). It can be used for research on acute lymphoblastic leukemia. It can also reduce MDM2 levels in HCT116 cells and upregulate p53 expression, indicating that it activates the p53 signaling pathway through USP7 inhibition. USP7-IN-3 can inhibit the proliferation of RS4;11 cells, demonstrating its high selectivity and efficacy against acute lymphoblastic leukemia cells.
  • Effective and selective allosteric inhibitor of ubiquitin-specific protease 7 (USP7).
  • Can be used for research on acute lymphoblastic leukemia.
  • Reduces MDM2 levels and upregulates p53 expression in HCT116 cells, activating the p53 signaling pathway through USP7 inhibition.
  • Inhibits proliferation of RS4;11 cells.
Catalog No. 50-004-36063 Supplier Medchemexpress LLC Supplier No. HY11212810MG
Add to Cart
Add to Cart
USP7-IN-3 (Compound 5) is an effective and selective allosteric inhibitor of ubiquitin-specific protease 7 (USP7). It can be used for research on acute lymphoblastic leukemia. It can also reduce MDM2 levels in HCT116 cells and upregulate p53 expression, indicating that it activates the p53 signaling pathway through USP7 inhibition. USP7-IN-3 can inhibit the proliferation of RS4;11 cells, demonstrating its high selectivity and efficacy against acute lymphoblastic leukemia cells.
  • Effective and selective allosteric inhibitor of ubiquitin-specific protease 7 (USP7).
  • Can be used for research on acute lymphoblastic leukemia.
  • Reduces MDM2 levels and upregulates p53 expression in HCT116 cells, activating the p53 signaling pathway through USP7 inhibition.
  • Inhibits proliferation of RS4;11 cells.