Learn More
Medchemexpress LLC Usp7-in-8 | 2009273-60-1 | 99.0% | C21H21N3O2 | 10MG
SDP
Catalog No. 5000260228
Encompass_Preferred
USP7-IN-8 is a selective small-molecule inhibitor of ubiquitin-specific protease 7 (USP7) used in biochemical and cellular research to probe deubiquitination pathways and investigate effects on protein stability, DNA repair, and cancer-related processes. It is supplied as a high-purity solid and is suitable for use in compound screening, biochemical assays, and cellular studies when handled under appropriate laboratory conditions.
  • Selective inhibition of USP7 with reported biochemical potency.
  • High chemical purity suitable for research applications.
  • Available in multiple solid and solution formats for experimental flexibility.
  • Stable under recommended storage conditions for extended shelf life.
  • Applicable to studies of protein ubiquitination, DNA repair, and cancer biology.
Catalog No. 50-002-60228 Supplier Medchemexpress LLC Supplier No. HY13481710MG
USP7-IN-8 is a selective small-molecule inhibitor of ubiquitin-specific protease 7 (USP7) used in biochemical and cellular research to probe deubiquitination pathways and investigate effects on protein stability, DNA repair, and cancer-related processes. It is supplied as a high-purity solid and is suitable for use in compound screening, biochemical assays, and cellular studies when handled under appropriate laboratory conditions.
  • Selective inhibition of USP7 with reported biochemical potency.
  • High chemical purity suitable for research applications.
  • Available in multiple solid and solution formats for experimental flexibility.
  • Stable under recommended storage conditions for extended shelf life.
  • Applicable to studies of protein ubiquitination, DNA repair, and cancer biology.