Learn More
Medchemexpress LLC USP7 ligand-linker conjugate 1 | 10 MG
SDP
Catalog No. 5000298400
Encompass_Preferred
USP7 Ligand-Linker Conjugates 1 is a ligand-linker conjugate intended as a building block for PROTAC synthesis that targets USP7. It links a USP7-binding ligand to a PROTAC-compatible linker to enable recruitment of E3 ligases and subsequent target degradation. Supplied for research use in medicinal chemistry and chemical biology under appropriate laboratory conditions.
  • Designed for PROTAC synthesis targeting USP7.
  • Combines a USP7-binding ligand with a PROTAC-compatible linker to recruit E3 ligases.
  • Molecular formula C38H49N5O6; molecular weight 671.83.
  • Supplied in small research quantities for chemical biology and medicinal chemistry.
  • Store pure material at -20°C for long-term stability; see datasheet for solvent storage guidance.
Catalog No. 50-002-98400 Supplier Medchemexpress LLC Supplier No. HY17536010MG
USP7 Ligand-Linker Conjugates 1 is a ligand-linker conjugate intended as a building block for PROTAC synthesis that targets USP7. It links a USP7-binding ligand to a PROTAC-compatible linker to enable recruitment of E3 ligases and subsequent target degradation. Supplied for research use in medicinal chemistry and chemical biology under appropriate laboratory conditions.
  • Designed for PROTAC synthesis targeting USP7.
  • Combines a USP7-binding ligand with a PROTAC-compatible linker to recruit E3 ligases.
  • Molecular formula C38H49N5O6; molecular weight 671.83.
  • Supplied in small research quantities for chemical biology and medicinal chemistry.
  • Store pure material at -20°C for long-term stability; see datasheet for solvent storage guidance.