Learn More
Medchemexpress LLC USP7 Ligand-Linker Conjugates 1 | 671.83 | 25MG
SDP
Catalog No. 5000383954
Encompass_Preferred
USP7 Ligand-Linker Conjugates 1 is a Target Protein Ligand-Linker Conjugate designed for the synthesis of PROTAC USP7 Degrader-1. This product incorporates a ligand for USP7 and a PROTAC linker, which recruits E3 ligases. PROTACs operate by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is intended for use as laboratory chemicals and in the manufacture of substances for research purposes only, to be handled by suitably qualified and experienced scientists in appropriately equipped and authorized facilities.
  • Incorporates a ligand for USP7
  • Recruits E3 ligases
  • Used for the synthesis of PROTAC USP7 Degrader-1
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-83954 Supplier Medchemexpress LLC Supplier No. 25MGHY175360
USP7 Ligand-Linker Conjugates 1 is a Target Protein Ligand-Linker Conjugate designed for the synthesis of PROTAC USP7 Degrader-1. This product incorporates a ligand for USP7 and a PROTAC linker, which recruits E3 ligases. PROTACs operate by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is intended for use as laboratory chemicals and in the manufacture of substances for research purposes only, to be handled by suitably qualified and experienced scientists in appropriately equipped and authorized facilities.
  • Incorporates a ligand for USP7
  • Recruits E3 ligases
  • Used for the synthesis of PROTAC USP7 Degrader-1
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins