Learn More
Medchemexpress LLC USP7 Ligand-Linker Conjugates 1 | 671.83 | 50 MG
SDP
Catalog No. 5000386276
Encompass_Preferred
USP7 Ligand-Linker Conjugates 1 is a Target Protein Ligand-Linker Conjugate that incorporates a ligand for USP7 and a PROTAC linker, which recruits E3 ligases. It can be used for the synthesis of PROTAC USP7 Degrader-1. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Incorporates a ligand for USP7
  • Includes a PROTAC linker which recruits E3 ligases
  • Can be used for the synthesis of PROTAC USP7 Degrader-1
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-86276 Supplier Medchemexpress LLC Supplier No. HY17536050MG
USP7 Ligand-Linker Conjugates 1 is a Target Protein Ligand-Linker Conjugate that incorporates a ligand for USP7 and a PROTAC linker, which recruits E3 ligases. It can be used for the synthesis of PROTAC USP7 Degrader-1. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Incorporates a ligand for USP7
  • Includes a PROTAC linker which recruits E3 ligases
  • Can be used for the synthesis of PROTAC USP7 Degrader-1
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins