Learn More
Medchemexpress LLC USP7 Ligand-Linker Conjugates 1 | 99.9% | 671.83 | 5 MG
SDP
Catalog No. 5000386380
Encompass_Preferred
USP7 Ligand-Linker Conjugates 1 is a Target Protein Ligand-Linker Conjugate that incorporates a ligand for USP7 and a PROTAC linker, which recruits E3 ligases. It can be used for the synthesis of PROTAC USP7 Degrader-1.
  • Target protein ligand-linker conjugates; deubiquitinase
  • Protac; cell cycle/DNA damage
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-86380 Supplier Medchemexpress LLC Supplier No. HY1753605MG
USP7 Ligand-Linker Conjugates 1 is a Target Protein Ligand-Linker Conjugate that incorporates a ligand for USP7 and a PROTAC linker, which recruits E3 ligases. It can be used for the synthesis of PROTAC USP7 Degrader-1.
  • Target protein ligand-linker conjugates; deubiquitinase
  • Protac; cell cycle/DNA damage
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins