Learn More
Cayman Chemical USP7-USP47 Inhibitor, Purity: ≥98%, CAS: 1247825-37-1, 5 mg
SDP
Catalog No. 502934312
Encompass_Preferred
A dual inhibitor of USP7 and USP47 (IC50s = 0.42 and 1 μM, respectively); selective for USP7 and USP47 over USP2, USP5, USP8, USP21, USP28, caspase-3, and cathepsin B (IC50s = >31.6 μM); inhibits the growth of HCT116 cells (EC50 = 7.6 μM)
Catalog No. 50-293-4312 Supplier Cayman Chemical Supplier No. 256225 MG
A dual inhibitor of USP7 and USP47 (IC50s = 0.42 and 1 μM, respectively); selective for USP7 and USP47 over USP2, USP5, USP8, USP21, USP28, caspase-3, and cathepsin B (IC50s = >31.6 μM); inhibits the growth of HCT116 cells (EC50 = 7.6 μM)