Learn More
Aviva Systems Biology Corp USP8 Antibody - N-terminal region (ARP59292_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59292_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against USP8. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human USP8. Alternate names: FLJ34456, HumORF8, KIAA0055, MGC129718, UBPYApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rat
Catalog No. 50-286-645