Learn More
Aviva Systems Biology Corp USP9X Antibody - C-terminal region (ARP59340_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59340_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against USP9X. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human USP9X. Alternate names: DFFRX, FAF, FAMApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-665