Learn More
Perkin Elmer US LLC USP/ICH Class 1 Elements + TI - Parenteral PDE
Catalog No. 502104170
Encompass_Preferred
Concentration of Element includes: Matrix Content Concentration 5% HNO3/2% HCl ICH-Class 1 Elements + TI Parental PDE As 15 µg/mL Tl 8 µg/mL Pb 5 µg/mL Hg 3 µg/mL Cd 2 µg/mL
Catalog No. 50-210-4170 Supplier Perkin Elmer US LLC Supplier No. N9304369
Concentration of Element includes: Matrix Content Concentration 5% HNO3/2% HCl ICH-Class 1 Elements + TI Parental PDE As 15 µg/mL Tl 8 µg/mL Pb 5 µg/mL Hg 3 µg/mL Cd 2 µg/mL