Learn More
Perkin Elmer US LLC USP/ICH Class 2A Elements – Oral and Parenteral PDEs
Catalog No. 502104167
Encompass_Preferred
Concentration of Element includes: Matrix Content Concentration 2% HNO3 ICH-Class 2A Elements Oral and Parenteral PDEs Ni 200 µg/mL V 100 µg/mL Co 50 µg/mL
Catalog No. 50-210-4167 Supplier Perkin Elmer US LLC Supplier No. N9304363
Concentration of Element includes: Matrix Content Concentration 2% HNO3 ICH-Class 2A Elements Oral and Parenteral PDEs Ni 200 µg/mL V 100 µg/mL Co 50 µg/mL