Learn More
Perkin Elmer US LLC USP/ICH Class 2B Precious Metal Elements (All PDEs)
Catalog No. 502104182
Encompass_Preferred
Concentration of Element includes: Matrix Concentration Part No 15% HCl N9304382 100 µg/mL: Os, Pd, Pt, Rh, Ru
Catalog No. 50-210-4182 Supplier Perkin Elmer US LLC Supplier No. N9304382
Concentration of Element includes: Matrix Concentration Part No 15% HCl N9304382 100 µg/mL: Os, Pd, Pt, Rh, Ru