Learn More
Perkin Elmer US LLC USP/ICH Class 2B Se & Ag - Parenteral PDE
Catalog No. 502104174
Encompass_Preferred
Concentration of Element includes: Matrix Content Concentration 2% HNO3 ICH-Class 2B Se and Ag Parenteral PDE Se 80 µg/mL Ag 10 µg/mL
Catalog No. 50-210-4174 Supplier Perkin Elmer US LLC Supplier No. N9304372
Concentration of Element includes: Matrix Content Concentration 2% HNO3 ICH-Class 2B Se and Ag Parenteral PDE Se 80 µg/mL Ag 10 µg/mL