Learn More
Perkin Elmer US LLC USP/ICH Class 3 Elements - Parenteral PDEs
Catalog No. 502104173
Encompass_Preferred
Concentration of Element includes: Matrix Content Concentration 2% HNO3/trace Tartaric Acid/ trace HF ICH-Class 3 Elements Parenteral PDEs Mo 150 µg/mL Cr 110 µg/mL Sn 60 µg/mL Cu 30 µg/mL Ba 70 µg/mL Li 25 µg/mL Sb 9 µg/mL
Catalog No. 50-210-4173 Supplier Perkin Elmer US LLC Supplier No. N9304373
Concentration of Element includes: Matrix Content Concentration 2% HNO3/trace Tartaric Acid/ trace HF ICH-Class 3 Elements Parenteral PDEs Mo 150 µg/mL Cr 110 µg/mL Sn 60 µg/mL Cu 30 µg/mL Ba 70 µg/mL Li 25 µg/mL Sb 9 µg/mL