Learn More
Perkin Elmer US LLC USP/ICH Elemental Impurities Standards Kit - Oral PDEs
Supplier:  Perkin Elmer US LLC N9304361
Encompass_Preferred
Concentration of Element includes: Matrix Concentration Part No 5% HNO3/2% HCl ICH-Class 1 Elements + TI Oral N9304362 30 µg/mL: Hg 15 µg/mL: As 8 µg/mL: Tl 5 µg/mL: Cd, Pb 2% HNO3 ICH-Class 2A Elements Oral and Parenteral PDEs N9304363 200 µg/mL: Ni 100 µg/mL: V 50 µg/mL: Co 2% HCl 100 µg/mL: Au N9303728 15% HCl ICH-Class 2BPrecious Metal Elements (no Os) All PDEs N9304364 100 µg/mL: lr, Pd, Pt, Rh, Ru 2% HNO3 ICH-Class 2B Se and Ag Oral PDE N9304365 150 µg/mL: Ag, Se 5% HNO3/trace Tartaric Acid/ trace HF ICH-Class 3 Elements Oral PDEs N9304366 1,100 µg/mL: Cr 600 µg/mL: Sn 300 µg/mL: Cu, Mo 140 µg/mL: Ba 120 µg/mL: Sb 55 µg/mL: Li 2% HNO3 Internal Standards All PDEs N9304367 50 µg/mL: Ge 20 µg/mL: Ga 1 µg/mL: ln, Tb
Catalog No. 50-210-4171