Learn More
Perkin Elmer US LLC USP/ICH Internal Standards - All PDEs
Supplier:  Perkin Elmer US LLC N9304367
Encompass_Preferred
Concentration of Element includes: Matrix Content Concentration 2% HNO3 Internal Standards All PDEs Ge 50 µg/mL Ga 20 µg/mL Ln, Tb 1 µg/mL
Catalog No. 50-210-4169