Description
- Entire Series Meets Military Ballistic High Velocity Standard
- MAX6™ Premium Anti-Fog Coating
- Sleek Co-Injected TPR over polycarbonate temples
- Fixed Closed Cell Foam Lined option
Certifications
CSA
Compliance
Meets ANSI Z87.1-2015, CSA Z94.3 and MIL - PRF - 31013 standards.
Specifications
Specifications
| Lens Coating | Anti-fog |
| Lens Tint | Gray |
| For Use With (Application) | Manufacturing |
| Frame Color | Black |
| No. per Pack | 1 Pair |
| Lens Material | Polycarbonate |
| Frame Material | Polycarbonate |