Learn More
World Precision Instrument Ussing System for Epithelial Research-USS7L
SDP
Supplier:  World Precision Instrument USS7L
Encompass
WPI’s classical Ussing Chambers are well established perfusion chambers that are easy to operate, easy to control temperature, and easy to clean after use. Ussing Cham­bers are machined from solid acrylic with eight entry ports for fluid lines, elec­trodes, or agar bridges. For easy, leak-free attachment of tubing and elec­trodes, all eight ports are Luer type. The four ports for volt­age and current electrodes are re­cessed to prevent formation of air bubbles in the chamber. The fluid com­part­ments in each side of the chamber are sep­a­rat­ed by the epithelial membrane being stud­ied. Sharp stainless steel pins on one side of the chamber hold the mem­brane in po­si­tion and mate with holes in the opposite chamber in­ter­face. (In the CHM4, tissue is held by an O-ring instead of pins.)
Catalog No. 50-252-1035