Learn More
Aviva Systems Biology Corp UST Antibody - C-terminal region (ARP46637_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46637_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against UST. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human UST. Alternate names: 2OSTApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53852