Learn More
Aviva Systems Biology Corp UST Antibody - N-terminal region (ARP46638_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46638_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UST. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human UST. Alternate names: 2OSTApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-268