Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters uStyragel GPC Column, 100000 A, 15-20 um Particle

Catalog No. 50785289 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This is a medium-efficiency GPC column designed for molecular weight screening. It features a polymer particle substrate and spherical particle shape, making it suitable for HPLC systems.

  • Chemistry: GPC
  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • pH range: 2 to 12
  • Molecular weight range: 50000 to 4000000
  • Particle shape: Spherical
  • Particle size: 15 to 20 μm
  • Pore size: 100000 Å
  • Format: Column
  • Packing solvent: Toluene
  • System: HPLC
  • Technique: GPC
  • Inner diameter: 7.8 mm
  • Length: 300 mm
  • Units per package: 1 unit

Catalog No. 50-785-289 Supplier Waters Corp Supplier No. WAT027164
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This is a medium-efficiency GPC column designed for molecular weight screening. It features a polymer particle substrate and spherical particle shape, making it suitable for HPLC systems.

  • Chemistry: GPC
  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • pH range: 2 to 12
  • Molecular weight range: 50000 to 4000000
  • Particle shape: Spherical
  • Particle size: 15 to 20 μm
  • Pore size: 100000 Å
  • Format: Column
  • Packing solvent: Toluene
  • System: HPLC
  • Technique: GPC
  • Inner diameter: 7.8 mm
  • Length: 300 mm
  • Units per package: 1 unit

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.