Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters GPC Column, μStyragel, 1,000,000Å, 15 to 20 μm, 7.8 × 300 mm

Catalog No. 50785288 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These medium-efficiency columns are designed for molecular weight screening.

  • Features GPC chemistry.
  • Supports SEC/GPC separation mode.
  • Constructed with polymer particle substrate.
  • Operates within a pH range of 2 to 12.
  • Provides a molecular weight range of 200,000 to 10,000,000.
  • Designed with spherical particle shape.
  • Particle size of 15 to 20 μm.
  • Features a pore size of 1,000,000 Å.
  • Packed in toluene solvent.
  • Compatible with HPLC systems.
  • Inner diameter of 7.8 mm.
  • Length of 300 mm.
  • Supplied as 1 unit per package.

Catalog No. 50-785-288 Supplier Waters Corp Supplier No. WAT027163
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These medium-efficiency columns are designed for molecular weight screening.

  • Features GPC chemistry.
  • Supports SEC/GPC separation mode.
  • Constructed with polymer particle substrate.
  • Operates within a pH range of 2 to 12.
  • Provides a molecular weight range of 200,000 to 10,000,000.
  • Designed with spherical particle shape.
  • Particle size of 15 to 20 μm.
  • Features a pore size of 1,000,000 Å.
  • Packed in toluene solvent.
  • Compatible with HPLC systems.
  • Inner diameter of 7.8 mm.
  • Length of 300 mm.
  • Supplied as 1 unit per package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.