Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp WATERS CORPORATION Column, 100Å, 15 to 20 μm, 7.8 x 300 mm

Catalog No. 50785292 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These are medium-efficiency columns specifically designed for molecular weight screening.

  • Chemistry: GPC
  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • pH range: 2 to 12
  • Molecular weight range: 50 to 1500
  • Particle shape: Spherical
  • Particle size: 15 to 20 μm
  • Pore size: 100 Å
  • Format: Column
  • Packing solvent: Toluene
  • System: HPLC
  • Technique: GPC
  • Inner diameter: 7.8 mm
  • Length: 300 mm
  • Units per package: 1 pk

Catalog No. 50-785-292 Supplier Waters Corp Supplier No. WAT027167
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These are medium-efficiency columns specifically designed for molecular weight screening.

  • Chemistry: GPC
  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • pH range: 2 to 12
  • Molecular weight range: 50 to 1500
  • Particle shape: Spherical
  • Particle size: 15 to 20 μm
  • Pore size: 100 Å
  • Format: Column
  • Packing solvent: Toluene
  • System: HPLC
  • Technique: GPC
  • Inner diameter: 7.8 mm
  • Length: 300 mm
  • Units per package: 1 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.