Learn More
Waters Corp Waters μStyragel GPC Column, 500 Å, 15 to 20 μm, 7.8 mm ID × 300 mm
Supplier:  Waters Corp WAT027161
Encompass_Preferred
This GPC column is designed for molecular weight screening applications, offering medium efficiency.
  • Medium-efficiency columns designed for molecular weight screening applications.
  • Contains hazardous material requiring special freight handling; additional charges may apply.
Catalog No. 50-785-287