Learn More
Medchemexpress LLC UT-11 | 99.6% | 400.32 | 5 MG
SDP
Catalog No. 5000359353
Encompass_Preferred
UT-11 is a potent, blood-brain barrier-permeable inhibitor of microsomal prostaglandin E synthase-1 (mPGES-1). It effectively inhibits PGE2 production in human and murine cells. Studies show UT-11 can dampen neuroinflammation by blunting the upregulation of mPGES-1 and other inflammatory genes in an LPS-induced inflammation mouse model.
  • Potent and blood-brain barrier-permeable inhibitor
  • Inhibits PGE2 production with IC50s of 0.10 μM in human (SK-N-AS) cells and 2.00 μM in murine (BV2) cells
  • Dampens neuroinflammation in LPS-induced inflammation mouse models
  • Blunts upregulation of mPGES-1 and inflammatory genes (IL-6, TNF-α, CCL2, CCL3, CCL4)
Catalog No. 50-003-59353 Supplier Medchemexpress LLC Supplier No. HY1492025MG
UT-11 is a potent, blood-brain barrier-permeable inhibitor of microsomal prostaglandin E synthase-1 (mPGES-1). It effectively inhibits PGE2 production in human and murine cells. Studies show UT-11 can dampen neuroinflammation by blunting the upregulation of mPGES-1 and other inflammatory genes in an LPS-induced inflammation mouse model.
  • Potent and blood-brain barrier-permeable inhibitor
  • Inhibits PGE2 production with IC50s of 0.10 μM in human (SK-N-AS) cells and 2.00 μM in murine (BV2) cells
  • Dampens neuroinflammation in LPS-induced inflammation mouse models
  • Blunts upregulation of mPGES-1 and inflammatory genes (IL-6, TNF-α, CCL2, CCL3, CCL4)