Learn More
Medchemexpress LLC HY-112895 5mg Medchemexpress, UT-155 CAS:2031161-35-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1128955MG
Encompass_Preferred
Medchemexpress, HY-112895 5mg UT-155 CAS:2031161-35-8 UT-155 is a selective and potent androgen receptor (AR) antagonist, with a Ki of 267 nM for UT-155 binding to AR-LBD. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9197