Learn More
Med Vet International Uterine Cytology Brush, J0273C, 29in, 10 Count
Catalog No. 5000583001
Encompass_Preferred
The use of a cytology brush is very common in female human patients. This cytology brush is a standard human cytology brush that is adhered to a culture swab insertion rod. It has the same double-guarded configuration as the JOR273. 10/box.
Catalog No. 50-005-83001 Supplier Med Vet International Supplier No. JOR273C
The use of a cytology brush is very common in female human patients. This cytology brush is a standard human cytology brush that is adhered to a culture swab insertion rod. It has the same double-guarded configuration as the JOR273. 10/box.