Learn More
Sino Biological Uteroglobin / SCGB1A1 Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 10808R030100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Uteroglobin / SCGB1A1 (rh Uteroglobin / SCGB1A1; Catalog#10808-H08H; P11684; Met1-Asn91).
Catalog No. 50-224-7478