Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories UTI Combo Strips

Catalog No. 23111293
Change view
Click to view available options
Quantity:
1 Test
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-293 1 Test
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-111-293 Supplier Germaine™ Laboratories Supplier No. 52301
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Germaine™ Laboratories UTI Combo Strips are urine dipsticks for the qualitative detection of Leukocytes and Nitrites

For OTC use, CLIA Waived

Specifications

Content And Storage 2°C to 30°C
Clia Complexity Waived
Detectable Analytes Leukocyte, Nitrite
Sample Type Urine
Disposable Single-use
For Use With (Application) Urinalysis
Quantity 1 Test
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.