Learn More
Medchemexpress LLC Utidelone (Epothilone D) | 189453-10-9 | C27H41NO5S | 50 MG
SDP
Catalog No. 5000373715
Encompass_Preferred
Utidelone (Epothilone D) is a potent microtubule stabilizer developed for research and analytical applications. It demonstrates significant cytotoxicity in human tumor cell lines, comparable to paclitaxel, and maintains its activity against multidrug-resistant cell lines. In vivo studies suggest it enhances microtubule density, axonal integrity, and cognitive functions.
  • Potent microtubule stabilizer
  • Exhibits cytotoxicity in human tumor cell lines
  • Effective against multidrug-resistant cell lines
  • Intended for research and analytical applications
  • Utilized in qualitative, quantitative, and methodological experiments
  • Supports HPLC, GC, and MS research
Catalog No. 50-003-73715 Supplier Medchemexpress LLC Supplier No. HY1527850MG
Utidelone (Epothilone D) is a potent microtubule stabilizer developed for research and analytical applications. It demonstrates significant cytotoxicity in human tumor cell lines, comparable to paclitaxel, and maintains its activity against multidrug-resistant cell lines. In vivo studies suggest it enhances microtubule density, axonal integrity, and cognitive functions.
  • Potent microtubule stabilizer
  • Exhibits cytotoxicity in human tumor cell lines
  • Effective against multidrug-resistant cell lines
  • Intended for research and analytical applications
  • Utilized in qualitative, quantitative, and methodological experiments
  • Supports HPLC, GC, and MS research