Learn More
Grainger JAMCO Utility Cart, Steel, 2,400 lb Capacity, 3 Shelves, 1 ea
Catalog No. 502788212
Encompass_Preferred
This heavy-duty steel utility cart is designed for efficient material transport in industrial, warehouse, and stockroom environments. It features an all-welded construction with three durable lipped and flush metal shelves, providing a stable 2,400 lb. total load capacity for demanding applications.
  • Features 12 gauge steel shelf construction with 1-1/2 in. lips for load retention.
  • Equipped with an ergonomic handle to improve operator comfort during transport.
  • Includes two rigid and two swivel 6 in. x 2 in. phenolic casters for smooth mobility.
  • Provides 12 in. of vertical clearance between shelves for organized storage of bulky items.
  • Finished with a resilient powder coating for increased durability and wear resistance.
  • Ships fully assembled for immediate deployment.
Catalog No. 50-278-8212 Supplier Grainger Supplier No. 16C039
This heavy-duty steel utility cart is designed for efficient material transport in industrial, warehouse, and stockroom environments. It features an all-welded construction with three durable lipped and flush metal shelves, providing a stable 2,400 lb. total load capacity for demanding applications.
  • Features 12 gauge steel shelf construction with 1-1/2 in. lips for load retention.
  • Equipped with an ergonomic handle to improve operator comfort during transport.
  • Includes two rigid and two swivel 6 in. x 2 in. phenolic casters for smooth mobility.
  • Provides 12 in. of vertical clearance between shelves for organized storage of bulky items.
  • Finished with a resilient powder coating for increased durability and wear resistance.
  • Ships fully assembled for immediate deployment.