Learn More
Grainger Jamco Utility Cart, 3,000 Lb. Capacity, Steel, 4 Shelves, 1 ea
Catalog No. 502791268
Encompass_Preferred
This heavy-duty utility cart features a robust welded steel construction, designed for efficient material handling in demanding industrial environments. It provides significant storage with four reinforced shelves, each capable of supporting substantial loads, and is finished with a durable powder coating for long-lasting use.
  • Capacity of 3,000 lbs. total load with 750 lbs. per shelf
  • Constructed from 12 ga. steel with 3/16 in. thick corner posts
  • Four-sided 1 1/2 in. lipped shelves to prevent load slippage
  • Equipped with 6 in. phenolic casters in a configuration of two rigid and two swivel for maneuverability
  • Ships fully assembled for immediate use
Catalog No. 50-279-1268 Supplier Grainger Supplier No. 16A937
This heavy-duty utility cart features a robust welded steel construction, designed for efficient material handling in demanding industrial environments. It provides significant storage with four reinforced shelves, each capable of supporting substantial loads, and is finished with a durable powder coating for long-lasting use.
  • Capacity of 3,000 lbs. total load with 750 lbs. per shelf
  • Constructed from 12 ga. steel with 3/16 in. thick corner posts
  • Four-sided 1 1/2 in. lipped shelves to prevent load slippage
  • Equipped with 6 in. phenolic casters in a configuration of two rigid and two swivel for maneuverability
  • Ships fully assembled for immediate use