Learn More
Grainger JAMCO Stock Cart, Welded Steel, 3,000 lb. Capacity, 6 Shelves, 1 ea.
Catalog No. 502791321
Encompass_Preferred
This stock cart is a robust material handling solution designed for industrial environments. Constructed from heavy-duty 12 ga. steel with a powder-coated finish, it provides durability and reliability for demanding tasks. The unit features a welded and bolted frame for structural integrity and includes six shelves, each with a 4-sided lip for secure transport of items.
  • Load capacity of 3,000 lbs. total
  • 500 lbs. capacity per shelf
  • 6 shelves with 1 1/2 in. lip height
  • Heavy-duty 6 in. phenolic casters in a (2) rigid and (2) swivel configuration
  • Overall dimensions of 54 in. x 31 in. x 72 in.
  • Flat handle design for easy maneuverability
Catalog No. 50-279-1321 Supplier Grainger Supplier No. 16A949
This stock cart is a robust material handling solution designed for industrial environments. Constructed from heavy-duty 12 ga. steel with a powder-coated finish, it provides durability and reliability for demanding tasks. The unit features a welded and bolted frame for structural integrity and includes six shelves, each with a 4-sided lip for secure transport of items.
  • Load capacity of 3,000 lbs. total
  • 500 lbs. capacity per shelf
  • 6 shelves with 1 1/2 in. lip height
  • Heavy-duty 6 in. phenolic casters in a (2) rigid and (2) swivel configuration
  • Overall dimensions of 54 in. x 31 in. x 72 in.
  • Flat handle design for easy maneuverability