Learn More
Grainger Jamco Stock Cart, Welded Steel, 3,000 Lb. Capacity, 5 Shelves, 1 ea
Catalog No. 502790763
Encompass_Preferred
This stock cart is a robust material handling solution designed for demanding environments. Constructed from durable 12 gauge welded steel with a protective powder coating, it provides reliable transport for heavy loads up to 3,000 lbs. The unit features five shelves with 1-1/2 in. lips to secure contents and is equipped with a comfortable ergonomic tubular handle for easy maneuvering.
  • Load capacity of 3,000 lbs. with a per-shelf capacity of 600 lbs.
  • Includes five shelves with 1-1/2 in. raised lips for product retention.
  • Durable welded steel construction with 12 gauge steel shelves and caster mounts.
  • Four 6 in. x 2 in. phenolic casters in a 2-rigid and 2-swivel configuration.
  • Ergonomic tubular handle for improved control and comfort during transport.
  • Overall dimensions of 42 in. x 25 in. x 68 in. with 13 in. shelf clearance.
Catalog No. 50-279-0763 Supplier Grainger Supplier No. 16C523
This stock cart is a robust material handling solution designed for demanding environments. Constructed from durable 12 gauge welded steel with a protective powder coating, it provides reliable transport for heavy loads up to 3,000 lbs. The unit features five shelves with 1-1/2 in. lips to secure contents and is equipped with a comfortable ergonomic tubular handle for easy maneuvering.
  • Load capacity of 3,000 lbs. with a per-shelf capacity of 600 lbs.
  • Includes five shelves with 1-1/2 in. raised lips for product retention.
  • Durable welded steel construction with 12 gauge steel shelves and caster mounts.
  • Four 6 in. x 2 in. phenolic casters in a 2-rigid and 2-swivel configuration.
  • Ergonomic tubular handle for improved control and comfort during transport.
  • Overall dimensions of 42 in. x 25 in. x 68 in. with 13 in. shelf clearance.