Learn More
Grainger Little Giant Utility Cart, Welded Steel, 2 Shelves, 3,600 lb. Capacity, 1 ea.
Catalog No. 502784253
Encompass_Preferred
This heavy-duty welded steel utility cart is designed for industrial material transport, offering a 3,600 lb total load capacity. The two-shelf unit features 4-sided lipped edges to secure items during transit and is finished with a durable powder coating for long-lasting performance.
  • Constructed from 12-gauge welded steel for heavy-duty applications
  • Two fixed shelves, each with a 1,800 lb capacity and 1.5 in. retaining lips
  • Equipped with four 8 in. non-marking polyurethane casters with a side pedal brake system
  • Caster configuration includes two rigid and two swivel casters for maneuverability
  • Includes a raised offset handle with a hand guard for ergonomic control
  • Industrial grade casters provide quiet operation and chemical resistance
  • Assembled unit measures 54 in. x 24 in. x 36 in.
Catalog No. 50-278-4253 Supplier Grainger Supplier No. 19G782
This heavy-duty welded steel utility cart is designed for industrial material transport, offering a 3,600 lb total load capacity. The two-shelf unit features 4-sided lipped edges to secure items during transit and is finished with a durable powder coating for long-lasting performance.
  • Constructed from 12-gauge welded steel for heavy-duty applications
  • Two fixed shelves, each with a 1,800 lb capacity and 1.5 in. retaining lips
  • Equipped with four 8 in. non-marking polyurethane casters with a side pedal brake system
  • Caster configuration includes two rigid and two swivel casters for maneuverability
  • Includes a raised offset handle with a hand guard for ergonomic control
  • Industrial grade casters provide quiet operation and chemical resistance
  • Assembled unit measures 54 in. x 24 in. x 36 in.