Learn More
Grainger 3M Utility Cloth Roll 314D, Aluminum Oxide, P120, 5/Case
Catalog No. 502760515
Encompass_Preferred
These utility cloth rolls are designed for versatile hand sanding applications, featuring a durable J-weight cloth backing and aluminum oxide abrasive. They are suitable for tasks ranging from deburring and blending to surface preparation and contaminant removal on metal surfaces.
  • Features P120 grit aluminum oxide abrasive for consistent finishing performance.
  • Constructed with a flexible J-weight cloth backing for conformability.
  • Closed coat construction provides a dense mineral coverage for effective material removal.
  • Ideal for maintenance, repair, and operations (MRO), marine, and metal fabrication industries.
  • Dimensions measure 1 in. (25.4 mm) in width by 50 yd. (45.72 m) in length.
  • Includes 5 rolls per case.
Catalog No. 50-276-0515 Supplier Grainger Supplier No. 163L96
These utility cloth rolls are designed for versatile hand sanding applications, featuring a durable J-weight cloth backing and aluminum oxide abrasive. They are suitable for tasks ranging from deburring and blending to surface preparation and contaminant removal on metal surfaces.
  • Features P120 grit aluminum oxide abrasive for consistent finishing performance.
  • Constructed with a flexible J-weight cloth backing for conformability.
  • Closed coat construction provides a dense mineral coverage for effective material removal.
  • Ideal for maintenance, repair, and operations (MRO), marine, and metal fabrication industries.
  • Dimensions measure 1 in. (25.4 mm) in width by 50 yd. (45.72 m) in length.
  • Includes 5 rolls per case.